Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium Chloride Conductivity Standard, 50,000 μS/cm (50 mS/cm) at 25°C, Ricca Chemical
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
Specifications
Packaging | Natural Poly Bottle |
Quantity | 500 mL |
Color | Colorless |
Assay Percent Range | 50, 000 ± 100 μS/cm at 25°C |
Odor | Odorless |
Solvent or Matrix | Aqueous Solution |
Physical Form | Liquid |
Chemical Name or Material | Potassium Chloride Conductivity Standard, 50, 000μS/cm (50, 000μmho/cm) at 25°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction