Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Chloride, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
1kg
2.5kg
500g
Packaging:
Poly Bottle

Chemical Identifiers

7447-40-7
ClK
74.548
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
4873
CHEBI:32588

High quality chemicals for laboratory and specialized industrial use

CAS 7447-40-7
Molecular Formula ClK
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
SMILES [Cl-].[K+]
IUPAC Name potassium;chloride
Molecular Weight (g/mol) 74.548
PubChem CID 4873
ChEBI CHEBI:32588
Formula Weight 74.55
Physical Form Crystals
Packaging Poly Bottle
Specific Gravity 1L = 1.99kg
Quantity 1kg
Chemical Name or Material Potassium Chloride
Grade Baker Analyzed™ ACS Reagent
Show More Show Less