Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Chloride, Crystal, Multi-Compendial, U.S.P., J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle
7447-40-7
ClK
74.548
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium chloride,enseal,muriate of potash,klotrix,sylvite,slow-k,potassium chloride kcl,klor-con,chlorvescent,kalitabs
4873
CHEBI:32588
potassium;chloride
[Cl-].[K+]
7447-40-7
ClK
74.548
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium chloride,enseal,muriate of potash,klotrix,sylvite,slow-k,potassium chloride kcl,klor-con,chlorvescent,kalitabs
4873
CHEBI:32588
potassium;chloride
[Cl-].[K+]
Formula Weight 74.55
Physical Form Crystals
Grade Multi-Compendial/USP
Packaging Poly Bottle
Specific Gravity 1L = 1.99kg
Quantity 500g
Chemical Name or Material Potassium Chloride