Alfa Aesar™ Potassium chloride crystal optic rectangle, 30mm x 15mm x 4mm, (drilled), polished both sides

Manufacturer: Alfa Aesar™ 38462KT

Quantity 1 each

 View more versions of this product

Catalog No. AA38462KT

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00011360
    Chemical Name or Material Potassium chloride crystal optic rectangle, 30mm x 15mm x 4mm, (drilled), polished both sides

    Specifications

    TSCA TSCA
    MDL Number MFCD00011360
    Quantity 1 each
    Chemical Name or Material Potassium chloride crystal optic rectangle, 30mm x 15mm x 4mm, (drilled), polished both sides

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit