Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Chloride, Crystal, Ultrapure Bioreagent, J.T.Baker™

Click to view available options
Quantity:
500g
5kg
Packaging:
Poly Bottle
Poly Pail
7447-40-7
ClK
74.548
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium chloride,enseal,muriate of potash,klotrix,sylvite,slow-k,potassium chloride kcl,klor-con,chlorvescent,kalitabs
4873
CHEBI:32588
potassium;chloride
[Cl-].[K+]
7447-40-7
ClK
74.548
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium chloride,enseal,muriate of potash,klotrix,sylvite,slow-k,potassium chloride kcl,klor-con,chlorvescent,kalitabs
4873
CHEBI:32588
potassium;chloride
[Cl-].[K+]

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For Molecular Biology Applications
Formula Weight 74.55
Physical Form Crystals
Grade Ultrapure Bioreagent
Packaging Poly Pail
Specific Gravity 1L = 1.99kg
Quantity 5kg
Chemical Name or Material Potassium Chloride