Potassium Chloride, Crystal, Ultrapure Bioreagent, J.T.Baker™

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

Manufacturer: J.T.Baker 400105

Packaging Poly Bottle
Quantity 2.5kg

 View more versions of this product

Catalog No. 02-003-740

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 7447-40-7
    Molecular Formula KCl
    Formula Weight 74.55
    Chemical Name or Material Potassium Chloride
    Grade Ultrapure Bioreagent

    Specifications

    Physical Form Crystals
    Formula Weight 74.55
    Packaging Poly Bottle
    Specific Gravity 1L = 1.99kg
    Quantity 2.5kg
    Molecular Formula KCl
    CAS 7447-40-7
    Chemical Name or Material Potassium Chloride
    Grade Ultrapure Bioreagent

    Description

      For Molecular Biology Applications

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit