Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Chloride, Crystal, USP, 99-100.5%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
45 kg
113.5 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7447-40-7
ClK
74.55
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium chloride
[Cl-].[K+]
7447-40-7
ClK
74.55
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium chloride
[Cl-].[K+]

PO165, 7447-40-7, KCl

Spectrum™ Potassium Chloride, Crystal, USP is used to treat hypokalemia. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Grade USP
CAS Max % 1
Percent Purity 99 to 100.5%
Bromide 0.001
Iodide 5e-005
Loss on Drying 0.01
Packaging Poly Pail
Quantity 12 kg