Potassium Chloroplatinate, 0.2% (w/v) in 10% (v/v) Hydrochloric Acid, Ricca Chemical

Manufacturer: Ricca Chemical CompanySupplier Diversity Partner 59504

Packaging Glass amber
Quantity 120mL

 View more versions of this product

Catalog No. 5950-4

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7732-18-5,7647-01-0,16921-30-5
    CAS Min % 87.0,10.32,0.18
    CAS Max % 91.0,10.75,0.18
    Physical Form Liquid
    Chemical Name or Material Potassium Chloroplatinate

    Specifications

    Color Clear light yellow
    Physical Form Liquid
    Quantity 120mL
    Concentration or Composition Notes 0.2% (w/v)
    Solvent or Matrix 10% (v/v) Hydrochloric Acid
    Packaging Glass amber
    CAS Max % 0.18,91.0,10.75
    CAS Min % 87.0,10.32,0.18
    CAS 16921-30-5,7732-18-5,7647-01-0
    Chemical Name or Material Potassium Chloroplatinate

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit