Potassium Chromate, 5% (w/v) Aqueous Solution, Ricca Chemical

Potassium Chromate, 5% (w/v) Aqueous Solution; Chloride free; Ricca Chemical; 500mL; Poly bottle; Indicator for argentometric titrations

Manufacturer: Ricca Chemical CompanySupplier Diversity Partner 60004

Packaging Plastic
Quantity 120mL

 View more versions of this product

Catalog No. 6000-4

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7732-18-5,7789-00-6,7761-88-8
    CAS Min % 93.0,4.67,7.0E-4
    CAS Max % 97.0,4.86,7.0E-4
    Chemical Name or Material Potassium Chromate

    Specifications

    Quantity 120mL
    CAS Max % 7.0E-4,97.0,4.86
    CAS 7732-18-5,7789-00-6,7761-88-8
    Concentration or Composition (by Analyte or Components) 5% (w/v) Aqueous Solution
    CAS Min % 93.0,4.67,7.0E-4
    Packaging Plastic
    Chemical Name or Material Potassium Chromate

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit