Potassium chromate, ACS, 99.0% min., Alfa Aesar™

For Use in Finishing Leather, Rust Proofing of Metals, Textile Mordant

Manufacturer: Alfa Aesar 1261036

Quantity 500g

 View more versions of this product

Catalog No. AA1261036

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7789-00-6
    Physical Form Crystalline
    Assay Percent Range 99.0% min.
    Molecular Formula K2CrO4
    Formula Weight 194.19
    MDL Number MFCD00011368
    Synonym Potassium chromium(VI) oxide
    Chemical Name or Material Potassium chromate, For analysis ACS
    Grade ACS Grade

    Specifications

    Formula Weight 194.19
    Quantity 500g
    Color White
    DOT Information DOT Class: 5.1, Packaging Group: III
    EINECS Number 232-140-5
    Physical Form Crystalline
    Odor Odorless
    Assay Percent Range 99.0% min.
    Molecular Formula K2CrO4
    Appearance Yellow crystals
    MDL Number MFCD00011368
    CAS 7789-00-6
    Vapor Pressure 0 hPa
    Health Hazard 1 H340-H350-H400-H410-H315-H319-H317-H335: May cause genetic defects. May cause cancer. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction. May cause respiratory irritation.
    Synonym Potassium chromium(VI) oxide
    Chemical Name or Material Potassium chromate, For analysis ACS
    Grade ACS Grade

    H340-H350-H400-H410-H315-H319-H317-H335: May cause genetic defects. May cause cancer. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction. May cause respiratory irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit