Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Chromate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle
7789-00-6
CrK2O4
194.189
XMXNVYPJWBTAHN-UHFFFAOYSA-N
potassium chromate,bipotassium chromate,potassium chromate vi,dipotassium monochromate,dipotassium chromate,tarapacaite,chromate of potash,chromate of potass,neutral potassium chromate,caswell no. 687
24597
CHEBI:75249
dipotassium;dioxido(dioxo)chromium
[O-][Cr](=O)(=O)[O-].[K+].[K+]
7789-00-6
CrK2O4
194.189
XMXNVYPJWBTAHN-UHFFFAOYSA-N
potassium chromate,bipotassium chromate,potassium chromate vi,dipotassium monochromate,dipotassium chromate,tarapacaite,chromate of potash,chromate of potass,neutral potassium chromate,caswell no. 687
24597
CHEBI:75249
dipotassium;dioxido(dioxo)chromium
[O-][Cr](=O)(=O)[O-].[K+].[K+]

High quality chemicals for laboratory and specialized industrial use

Formula Weight 194.19
Physical Form Crystals
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 2.73kg
Quantity 2.5kg
Chemical Name or Material Potassium Chromate