Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Chromate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
125g
500g
2.5kg
Packaging:
Poly Bottle

Chemical Identifiers

7789-00-6
CrK2O4
194.189
XMXNVYPJWBTAHN-UHFFFAOYSA-N
24597
CHEBI:75249

High quality chemicals for laboratory and specialized industrial use

SMILES [O-][Cr](=O)(=O)[O-].[K+].[K+]
IUPAC Name dipotassium;dioxido(dioxo)chromium
Formula Weight 194.19
Physical Form Crystals
Packaging Poly Bottle
Specific Gravity 1L = 2.73kg
Quantity 125g
Chemical Name or Material Potassium Chromate
Grade Baker Analyzed™ ACS Reagent