Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Citrate Monohydrate (Crystalline/Certified), Fisher Chemical

Click to view available options
Quantity:
500g
Packaging:
Glass Bottle
6100-05-6
C6H7K3O8
324.41
MFCD00150442
PJAHUDTUZRZBKM-UHFFFAOYSA-K
potassium citrate monohydrate, tripotassium citrate monohydrate, unii-ee90oni6ff, ee90oni6ff, citric acid tripotassium salt, urocit-k, ccris 6543, tri-potassium citrate monohydrate, potassium citrate tribasic monohydrate, citric acid, tripotassium salt, monohydrate
2735208
CHEBI:64746
tripotassium;2-hydroxypropane-1,2,3-tricarboxylate;hydrate
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.[K+].[K+].[K+]
6100-05-6
C6H7K3O8
324.41
MFCD00150442
PJAHUDTUZRZBKM-UHFFFAOYSA-K
potassium citrate monohydrate, tripotassium citrate monohydrate, unii-ee90oni6ff, ee90oni6ff, citric acid tripotassium salt, urocit-k, ccris 6543, tri-potassium citrate monohydrate, potassium citrate tribasic monohydrate, citric acid, tripotassium salt, monohydrate
2735208
CHEBI:64746
tripotassium;2-hydroxypropane-1,2,3-tricarboxylate;hydrate
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.[K+].[K+].[K+]
Packaging Glass Bottle
Melting Point 180°C
pH 7.5 to 9.0
Quantity 500g
Identification Pass Test
Color White
Linear Formula KO2CCH2C(OH)(CO2K)CH2CO2K·H2O
Solubility Information 0.01% max. Insoluble Matter
Formula Weight 324.41
Physical Form Solid
Grade Certified
Chemical Name or Material Potassium Citrate Monohydrate
Show More Show Less