Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Citrate, Monohydrate, Granular, FCC, 99-100.5%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6100-05-6
C6H7K3O8
324.41
PJAHUDTUZRZBKM-UHFFFAOYSA-K
tripotassium 2-hydroxypropane-1,2,3-tricarboxylate hydrate
O.[K+].[K+].[K+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
6100-05-6
C6H7K3O8
324.41
PJAHUDTUZRZBKM-UHFFFAOYSA-K
tripotassium 2-hydroxypropane-1,2,3-tricarboxylate hydrate
O.[K+].[K+].[K+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

P1266, 6100-05-6, C6H5K3O7•H2O

Spectrum™ Potassium Citrate, Monohydrate, Granular, FCC is used in foods and beverages as buffering, sequestering or emulsifying agent. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Grade FCC
CAS Max % 1
Percent Purity 99 to 100.5%
Lead (Pb) 2 mg/kg
Loss on Drying 3-6%
Packaging Fiber Drum
Quantity 50 kg