Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Citrate, Monohydrate, Granular, U.S.P. - F.C.C., J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Wide Mouth Amber Glass Bottle

Chemical Identifiers

6100-05-6
C6H7K3O8
324.41
PJAHUDTUZRZBKM-UHFFFAOYSA-K
2735208
CHEBI:64746
tripotassium;2-hydroxypropane-1,2,3-tricarboxylate;hydrate
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.[K+].[K+].[K+]
Formula Weight 324.42
Physical Form Granular
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 1.98kg
Quantity 2.5kg
Chemical Name or Material Potassium Citrate, Monohydrate
Grade FCC/USP