Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Citrate, Monohydrate, Granular, USP, 99-100.5%, Spectrum™

Click to view available options
Quantity:
50 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6100-05-6
C6H7K3O8
324.41
PJAHUDTUZRZBKM-UHFFFAOYSA-K
tripotassium 2-hydroxypropane-1,2,3-tricarboxylate hydrate
O.[K+].[K+].[K+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
6100-05-6
C6H7K3O8
324.41
PJAHUDTUZRZBKM-UHFFFAOYSA-K
tripotassium 2-hydroxypropane-1,2,3-tricarboxylate hydrate
O.[K+].[K+].[K+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

PO175, 6100-05-6, C6H5K3O7•H2O

Spectrum™ Potassium Citrate, Monohydrate, Granular, USP belongs to a class of drugs known as urinary alkalinizers. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Grade USP
CAS Max % 1
Percent Purity 99 to 100.5%
Loss on Drying 3-6%
Packaging Poly Bottle
Quantity 500 g