Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium cyanide, ACS, 96.0% min., Alfa Aesar™

Click to view available options
Quantity:
50g
250g
1kg
151-50-8
CKN
65.116
MFCD00011397
NNFCIKHAZHQZJG-UHFFFAOYSA-N
potassium cyanide,potassium cyanide k cn,cyanide of potassium,caswell no. 688a,zyankali,kalium cyanid german,unii-mqd255m2zo,rcra waste number p098,cyanure de potassium french
9032
potassium;cyanide
[C-]#N.[K+]
151-50-8
CKN
65.116
MFCD00011397
NNFCIKHAZHQZJG-UHFFFAOYSA-N
potassium cyanide,potassium cyanide k cn,cyanide of potassium,caswell no. 688a,zyankali,kalium cyanid german,unii-mqd255m2zo,rcra waste number p098,cyanure de potassium french
9032
potassium;cyanide
[C-]#N.[K+]
Assay Percent Range 96.0% min.
Beilstein 4652394
Solubility Information Highly soluble in water. Soluble in methanol,glycerol,and formamide. Slightly soluble in ethanol.
Formula Weight 65.12
Physical Form Granular
Grade ACS Reagent
Density 1.52 g/mL
Sensitivity Hygroscopic
Boiling Point 1625°C
Melting Point 634°C
Quantity 250g
Chemical Name or Material Potassium cyanide, For analysis ACS
Show More Show Less

P234-P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P390-P501c

H290-H300+H310+H330-H372

DOTInformation : Hazard Class: 6.1; Packaging Group: I

EINECSNumber : 205-792-3

RTECSNumber : TS8750000

TSCA : Yes

Recommended Storage : Ambient temperatures