Alfa Aesar™ Potassium cyclohexanebutyrate, AAS, K 18.8%

Manufacturer: Alfa Aesar™ 8726506

Quantity 5g

 View more versions of this product

Catalog No. AA8726506

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 62638-03-3
    Molecular Formula C10H17KO2
    Formula Weight 208.33
    MDL Number MFCD00013051
    Synonym Cyclohexanebutyric acid potassium salt; Potassium 4-cyclohexylbutyrate
    Chemical Name or Material Potassium cyclohexanebutyrate
    Grade AAS Grade

    Specifications

    Formula Weight 208.33
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Health Hazard 1 Warning
    Color White
    Physical Form Powder
    MDL Number MFCD00013051
    Concentration K 18.8%
    Quantity 5g
    Sensitivity Hygroscopic
    Molecular Formula C10H17KO2
    CAS 62638-03-3
    Synonym Cyclohexanebutyric acid potassium salt; Potassium 4-cyclohexylbutyrate
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material Potassium cyclohexanebutyrate
    Grade AAS Grade

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit