Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Potassium D-gluconate, 99%
Click to view available options
Quantity:
50g
250g
1000g
Description
Applications
Potassium D-gluconate widely used as sequestering compounds for metal ions in alkaline systems. It is used in a newly developed process entitled ARS for the prevention of shrinkage in dyed wool fabric. Potassium gluconate has been shown to elevate lactate production, and ATP content, as well as induce the Hemoglobin (methemoglobin) reduction rate in rat erythrocytes. It is used in cell culture and in large scale applications such as the cleaning of metal surfaces.
Solubility
Soluble in water. (50 mg/ml).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
Potassium D-gluconate widely used as sequestering compounds for metal ions in alkaline systems. It is used in a newly developed process entitled ARS for the prevention of shrinkage in dyed wool fabric. Potassium gluconate has been shown to elevate lactate production, and ATP content, as well as induce the Hemoglobin (methemoglobin) reduction rate in rat erythrocytes. It is used in cell culture and in large scale applications such as the cleaning of metal surfaces.
Solubility
Soluble in water. (50 mg/ml).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
Specifications
Solubility Information | Soluble in water. (50mg/ml). |
Formula Weight | 234.25 |
Percent Purity | 99% |
Melting Point | ~180°C (decomposition) |
Quantity | 50g |
Chemical Name or Material | Potassium D-gluconate |
Safety and Handling
EINECSNumber : 206-074-2
RTECSNumber : LZ5230000
TSCA : Yes
Recommended Storage : Ambient temperatures