Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Potassium D-gluconate, 99%

Click to view available options
Quantity:
50g
250g
1000g
299-27-4
C6H11KO7
234.245
MFCD00064211
HLCFGWHYROZGBI-JJKGCWMISA-M
potassium gluconate,potassium d-gluconate,gluconic acid potassium salt,potassuril,kaon elixir,potassium 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanoate,katorin,potalium,potasoral,sirokal
16760467
potassium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[K+]
299-27-4
C6H11KO7
234.245
MFCD00064211
HLCFGWHYROZGBI-JJKGCWMISA-M
potassium gluconate,potassium d-gluconate,gluconic acid potassium salt,potassuril,kaon elixir,potassium 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanoate,katorin,potalium,potasoral,sirokal
16760467
potassium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[K+]
Applications
Potassium D-gluconate widely used as sequestering compounds for metal ions in alkaline systems. It is used in a newly developed process entitled ARS for the prevention of shrinkage in dyed wool fabric. Potassium gluconate has been shown to elevate lactate production, and ATP content, as well as induce the Hemoglobin (methemoglobin) reduction rate in rat erythrocytes. It is used in cell culture and in large scale applications such as the cleaning of metal surfaces.

Solubility
Soluble in water. (50 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
Formula Weight 234.25
Percent Purity 99%
Melting Point ~180°C (decomposition)
Quantity 250g
Chemical Name or Material Potassium D-gluconate

RTECSNumber : LZ5230000

TSCA : TSCA