Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Dichromate, 0.025N, ASTM/EPA, Ricca Chemical

Manufacturer:  Ricca Chemical CompanySupplier Diversity Partner 605032

Packaging Plastic
Quantity 1L

 View more versions of this product

Catalog No. 6050-32


 

Due to product restrictions, please to purchase or view availability for this product.


Specifications Description & Specifications

Chemical Identifiers

Potassium Dichromate

Specifications

Plastic
1L
Potassium Dichromate
ASTM/EPA for Chemical Oxygen Demand (COD)
0.0250N (N/40)
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit