Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium dichromate, 0.25N Standardized Solution, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 mL
1 L
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Orange |
Linear Formula | K2Cr2O7 |
UN Number | UN3287 |
Concentration | 0.25N |
Formula Weight | 294.18 |
Odor | Odorless |
Quantity | 1 L |
Physical Form | Liquid |
Chemical Name or Material | Potassium dichromate, Standardized Solution |
Safety and Handling
Hazard Category | H315-H317-H319-H332-H334-H340-H350-H360FD-H373 |
Hazard Statement | GHS H Statement H272-H301-H331-H334-H340-H350-H360-H372-H314-H312-H317 May intensify fire oxidizer. Toxic if swallowed. Toxic if inhaled. May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause genetic defects. May cause cancer. May damage fertility or the unborn child. Causes damage to organs through prolonged or repeated exposure. Causes severe skin burns and eye damage. Harmful in contact with skin. May cause an allergic skin reaction. |
Precautionary Statement | P201-P202-P260-P264b-P271-P272-P280-P281-P285-P302+P352-P304+P340-P305+P351+P338-P308+P313-P333+P313-P342+P311-P362-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC LIQUID, INORGANIC, N.O.S. |
EINECSNumber | 231-906-6 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction