Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Potassium dichromate, 0.25N Standardized Solution

Manufacturer:  Alfa Aesar™ 35587K7

Quantity 4L

 View more versions of this product

Catalog No. AA35587K7


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7778-50-9
294.18
UN3287
K2Cr2O7
MFCD00011367
Potassium dichromate, Standardized Solution

Specifications

7778-50-9
0.25N
294.18
UN3287
Orange
Potassium dichromate, Standardized Solution
Liquid
K2Cr2O7
MFCD00011367
Odorless
4L

Safety and Handling

SDS & Safety Documents

GHS H Statement
H272-H301-H331-H334-H340-H350-H360-H372-H314-H312-H317
May intensify fire
oxidizer.
Toxic if swallowed.
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause genetic defects.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
May cause an allergic skin reaction.

GHS P Statement
P221-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC LIQUID, INORGANIC, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit