Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium dichromate, 99.5%, Acros Organics™

Click to view available options
Quantity:
500g
1kg
5kg
Packaging:
Plastic bottle
7778-50-9
Cr2K2O7
294.19
MFCD00011367
KMUONIBRACKNSN-UHFFFAOYSA-N
potassium dichromate,potassium bichromate,kaliumdichromat,iopezite,dipotassium bichromate,dipotassium dichromate,potassium dichromate vi,dichromic acid dipotassium salt,dipotassium dichromium heptaoxide,bichromate of potash
24502
CHEBI:53444
dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]
7778-50-9
Cr2K2O7
294.19
MFCD00011367
KMUONIBRACKNSN-UHFFFAOYSA-N
potassium dichromate,potassium bichromate,kaliumdichromat,iopezite,dipotassium bichromate,dipotassium dichromate,potassium dichromate vi,dichromic acid dipotassium salt,dipotassium dichromium heptaoxide,bichromate of potash
24502
CHEBI:53444
dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]

Oxidation reagent

Oxidation reagent
Assay Percent Range 99.5%
Linear Formula K2Cr2O7
Fieser 08,410; 09,383; 10,324; 12,405
Merck Index 15, 7748
Solubility Information Solubility in water: 125g/L (20°C). Other solubilities: 50 g/100 mL water (100°C), insoluble in alcohol
Formula Weight 294.19
Physical Form Crystals
Percent Purity 99.5%
Grade Extra Pure
Chlorine (Cl) 700ppm max.
Insoluble in Water 0.05% max.
Loss on Drying 0.05% max. (110°C)
Packaging Plastic bottle
Solubility Solubility in water: 125g/L (20°C). Other solubilities: 50 g/100 mL water (100°C), insoluble in alcohol
Sulfate 300ppm max.
Boiling Point 500.0°C
Color Orange to Orange-Red
Melting Point 398.0°C
Quantity 5kg
Chemical Name or Material Potassium dichromate
Show More Show Less

GHS H Statement
May cause an allergic skin reaction.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause genetic defects.
May cause cancer.
Fatal in contact with skin.
Causes damage to organs through prolonged or repeated exposure.
May damage fertility. May damage the unborn child.
Toxic if swallowed.
May intensify fire; oxidizer.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Take any precaution to avoid mixing with combustibles.
In case of major fire and large quantities: Evacuate area. Fight fire remotely due to the risk of explosion.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

Danger