Alfa Aesar™ Potassium dichromate, Acculute Standard Volumetric Solution, Final Concentration 0.1N

Manufacturer: Alfa Aesar™ 88598LS

Quantity 6 units

 View more versions of this product

Catalog No. AA88598LS

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 7778-50-9
    Molecular Formula Cr2K2O7
    Linear Formula K2Cr2O7
    Formula Weight 294.18
    MDL Number MFCD00011367
    UN Number UN3287
    Chemical Name or Material Potassium dichromate Acculute Standard Volumetric Solution, Final Concentration 0.1N

    Specifications

    Formula Weight 294.18
    Health Hazard 3 GHS P Statement
    P221-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H272-H301-H331-H334-H340-H350-H360-H372-H314-H312-H317
    May intensify fire
    oxidizer.
    Toxic if swallowed.
    Toxic if inhaled.
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause genetic defects.
    May cause cancer.
    May damage fertility or the unborn child.
    Causes damage to organs through prolonged or repeated exposure.
    Causes severe skin burns and eye damage.
    Harmful in contact with skin.
    May cause an allergic skin reaction.
    Linear Formula K2Cr2O7
    Quantity 6 units
    Physical Form Liquid
    UN Number UN3287
    Color Orange
    MDL Number MFCD00011367
    CAS 7778-50-9
    Health Hazard 1 Danger
    Odor Odorless
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC LIQUID, INORGANIC, N.O.S.
    TSCA TSCA
    Molecular Formula Cr2K2O7
    Chemical Name or Material Potassium dichromate Acculute Standard Volumetric Solution, Final Concentration 0.1N

    GHS H Statement
    H272-H301-H331-H334-H340-H350-H360-H372-H314-H312-H317
    May intensify fire
    oxidizer.
    Toxic if swallowed.
    Toxic if inhaled.
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause genetic defects.
    May cause cancer.
    May damage fertility or the unborn child.
    Causes damage to organs through prolonged or repeated exposure.
    Causes severe skin burns and eye damage.
    Harmful in contact with skin.
    May cause an allergic skin reaction.

    GHS P Statement
    P221-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit