Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Dichromate, ACROS Organics™

Click to view available options
Quantity:
1L
7778-50-9,7732-18-5
Cr2K2O7
294.182
MFCD00011367
KMUONIBRACKNSN-UHFFFAOYSA-N
potassium dichromate,potassium bichromate,kaliumdichromat,iopezite,dipotassium bichromate,dipotassium dichromate,potassium dichromate vi,dichromic acid dipotassium salt,dipotassium dichromium heptaoxide,bichromate of potash
24502
CHEBI:53444
dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]
7778-50-9,7732-18-5
Cr2K2O7
294.182
MFCD00011367
KMUONIBRACKNSN-UHFFFAOYSA-N
potassium dichromate,potassium bichromate,kaliumdichromat,iopezite,dipotassium bichromate,dipotassium dichromate,potassium dichromate vi,dichromic acid dipotassium salt,dipotassium dichromium heptaoxide,bichromate of potash
24502
CHEBI:53444
dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]
Merck Index 15, 7748
Formula Weight 294.17
Grade Extra Pure
Density 1.01g/mL
Quantity 1L
Chemical Name or Material Potassium dichromate

GHS H Statement:
May cause genetic defects.
Toxic to aquatic life with long lasting effects.
May cause cancer.
Harmful if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Causes serious eye damage.
Causes skin irritation.
May cause an allergic sk

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger