Potassium dichromate, ACS, 99.0% min, Alfa Aesar™

For Use As An Oxidizing Agent, in Dyeing, Painting

Manufacturer: Alfa Aesar 1345036

Quantity 500g

 View more versions of this product

Catalog No. AA1345036

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 7778-50-9
    Physical Form Crystalline Powder
    Assay Percent Range 99.0% min.
    Molecular Formula Cr2K2O7
    Formula Weight 294.18
    MDL Number MFCD00011367
    Synonym Potassium bichromate
    Chemical Name or Material Potassium dichromate, For ACS analysis
    Grade ACS Grade

    Specifications

    Physical Form Crystalline Powder
    Formula Weight 294.18
    Quantity 500g
    Color Orange
    DOT Information Hazard Class: 5.1, Packing Group: II
    Density 2.676g/cm3
    EINECS Number 231-906-6
    Assay Percent Range 99.0% min.
    Molecular Formula Cr2K2O7
    Appearance Orange crystalline powder
    MDL Number MFCD00011367
    CAS 7778-50-9
    Synonym Potassium bichromate
    Health Hazard 1 H272-H301-H330-H312-H317-H334-H340-H350-H360-H372-H314-H400-H410: May intensify fire; oxidizer. Toxic if swallowed. Fatal if inhaled. Harmful in contact with skin. May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause genetic defects. May cause cancer. May damage fertility or the unborn child. Causes damage to organs through prolonged or repeated exposure. Causes severe skin burns and eye damage. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
    Chemical Name or Material Potassium dichromate, For ACS analysis
    Grade ACS Grade

    H272-H301-H330-H312-H317-H334-H340-H350-H360-H372-H314-H400-H410: May intensify fire; oxidizer. Toxic if swallowed. Fatal if inhaled. Harmful in contact with skin. May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause genetic defects. May cause cancer. May damage fertility or the unborn child. Causes damage to organs through prolonged or repeated exposure. Causes severe skin burns and eye damage. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit