Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Dichromate, ACS, Reagents

Catalog No. 01337851
Click to view available options
Quantity:
100 g
500 g
2.5 kg

Potassium Dichromate, ACS, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7778-50-9
Molecular Formula Cr2K2O7
Molecular Weight (g/mol) 294.18
InChI Key KMUONIBRACKNSN-UHFFFAOYSA-N
Synonym Potassium bichromate
IUPAC Name dipotassium [(oxidodioxochromio)oxy]chromiumoylolate
SMILES [K+].[K+].[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O

Specifications

Boiling Point 500°C
Melting Point 398°C
Color Orange/Red
Physical Form Solid
Quantity 2.5 kg
Grade ACS
Packaging Poly Bottle
Specific Gravity 2.676
Chemical Name or Material Potassium Dichromate, ACS
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.