Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Dichromate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
2.5kg
500g
Packaging:
Poly Bottle

Chemical Identifiers

7778-50-9
Cr2K2O7
294.182
KMUONIBRACKNSN-UHFFFAOYSA-N
24502
CHEBI:53444

High quality chemicals for laboratory and specialized industrial use

CAS 7778-50-9
Molecular Formula Cr2K2O7
InChI Key KMUONIBRACKNSN-UHFFFAOYSA-N
SMILES [O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]
IUPAC Name dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
Molecular Weight (g/mol) 294.182
PubChem CID 24502
ChEBI CHEBI:53444
Formula Weight 294.19
Physical Form Crystals
Packaging Poly Bottle
Specific Gravity 1L = 2.68kg
Quantity 500g
Chemical Name or Material Potassium Dichromate
Grade Baker Analyzed™ ACS Reagent
Show More Show Less