Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Dichromate Solution, 0.02M, Honeywell Fluka™

Click to view available options
Quantity:
1L
6 x 1L
7778-50-9
Cr2K2O7
294.182
MFCD00011367
KMUONIBRACKNSN-UHFFFAOYSA-N
potassium dichromate,potassium bichromate,kaliumdichromat,iopezite,dipotassium bichromate,dipotassium dichromate,potassium dichromate vi,dichromic acid dipotassium salt,dipotassium dichromium heptaoxide,bichromate of potash
24502
CHEBI:53444
dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]
7778-50-9
Cr2K2O7
294.182
MFCD00011367
KMUONIBRACKNSN-UHFFFAOYSA-N
potassium dichromate,potassium bichromate,kaliumdichromat,iopezite,dipotassium bichromate,dipotassium dichromate,potassium dichromate vi,dichromic acid dipotassium salt,dipotassium dichromium heptaoxide,bichromate of potash
24502
CHEBI:53444
dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]

Volumetric, 0.02 M K2Cr2O7, for COD determination according to DIN 38409, part 41

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

Linear Formula K2Cr2O7
UN Number NONH for all modes of transport
Formula Weight 294.18g/mol
Quantity 1L
Chemical Name or Material Potassium dichromate solution

P201-P273-P308 + P313-P391-P501