Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium Dichromate Spectrophotometric Standard, 1.8 AU/cm at 280 nm, Ricca Chemical
Click to view available options
Quantity:
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
Specifications
Packaging | Cubitainer |
Quantity | 20 L |
CAS Min % | 98.0, 0.02 |
CAS Max % | 100.0, 0.02 |
Color | Orange |
Physical Form | Liquid |
Chemical Name or Material | Potassium Dichromate Spectrophotometric Standard, 1.8AU/cm at 280nm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction