Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Dichromate TS, (U.S.P. Test Solution), Spectrum™

Click to view available options
Quantity:
100 mL
1 L
Packaging:
Amber Glass Bottle
7778-50-9,7732-18-5
Cr2K2O7
294.18
KMUONIBRACKNSN-UHFFFAOYSA-N
dipotassium [(oxidodioxochromio)oxy]chromiumoylolate
[K+].[K+].[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O
7778-50-9,7732-18-5
Cr2K2O7
294.18
KMUONIBRACKNSN-UHFFFAOYSA-N
dipotassium [(oxidodioxochromio)oxy]chromiumoylolate
[K+].[K+].[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O

P-226, 7778-50-9

Spectrum™ Potassium Dichromate TS, (U.S.P. Test Solution) is a solution, sometimes designated with "TS or Test Solution", that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.

CAS Max % 0.001,0.999
UN Number UN3287
Grade Chemical Solution
Packaging Amber Glass Bottle
Quantity 1 L