Alfa Aesar™ Potassium dihydrogen citrate hydrate, 99% (dry basis)

Manufacturer: Alfa Aesar™ 4441536

Quantity 500g

 View more versions of this product

Catalog No. AA4441536

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 866-83-1
    Molecular Formula HOOCCH2C(OH)(COOH)CH2COOK·xH2O
    Formula Weight 230.22 (Anhydrous)
    MDL Number MFCD00036472
    Beilstein 4772973
    Merck Index 14,7624
    Synonym Potassium citrate monobasic
    Chemical Name or Material Potassium dihydrogen citrate hydrate

    Specifications

    Physical Form Powder/Chunks
    Formula Weight 230.22 (Anhydrous)
    Quantity 500g
    TSCA TSCA
    Molecular Formula HOOCCH2C(OH)(COOH)CH2COOK·xH2O
    CAS 866-83-1
    Merck Index 14,7624
    Sensitivity Hygroscopic
    MDL Number MFCD00036472
    Beilstein 4772973
    Synonym Potassium citrate monobasic
    Chemical Name or Material Potassium dihydrogen citrate hydrate
    Assay 99%, (dry basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit