Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium dihydrogen phosphate, 98+%, Alfa Aesar™

Manufacturer:  Alfa Aesar A121420C

Quantity 10000g

 View more versions of this product

Catalog No. AAA121420C


  Check Availability
Add to cart

Specifications

Description

Applications
Potassium dihydrogen phosphate is used as a food additive, fertilizer (NPK), paint additive, coating additive, adhesive, buffering agent and in optical modulators. It is extensively used in research and analytical laboratories as a buffer in mobile/sample preparations for analysis by high pressure liquid chromatography (HPLC) and also in organic synthesis. MKP, monopotassium phosphate, owing to its non-linear optical properties, is used in optical modulators and for second-harmonic generation (SHG).

Solubility
Soluble in water. Slightly soluble in ethanol.

Notes
Loses H{2}O at 400°, forming metaphosphateHygroscopic.
Description & Specifications

Chemical Identifiers

7778-77-0
KH2PO4
MFCD00011401
Potassium phosphate, monobasic
H2KO4P
136.09
14,7659
Potassium dihydrogen phosphate

Specifications

7778-77-0
KH2PO4
MFCD00011401
Potassium phosphate, monobasic
10000g
≥98%
H2KO4P
136.09
14,7659
253°C
Potassium dihydrogen phosphate

Safety and Handling

SDS & Safety Documents

GHS H Statement
H315-H319;
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313;
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse. Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

RTECSNumber : TC6615500

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit