Potassium dihydrogen phosphate, ACS, 99.0% min., Alfa Aesar™

For Use in Buffers For pH Determination

Manufacturer: Alfa Aesar 11594A1

Quantity 1kg

 View more versions of this product

Catalog No. AA11594A1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7778-77-0
    Physical Form Crystalline
    Assay Percent Range 99.0% min.
    Molecular Formula H2KO4P
    Formula Weight 136.09
    MDL Number MFCD00011401
    Synonym Potassium orthophosphate
    Chemical Name or Material Potassium dihydrogen phosphate, For analysis ACS
    Grade ACS Grade

    Specifications

    Formula Weight 136.09
    Quantity 1kg
    Color White
    EINECS Number 231-913-4
    Physical Form Crystalline
    Assay Percent Range 99.0% min.
    Molecular Formula H2KO4P
    Synonym Potassium orthophosphate
    Appearance White crystals
    MDL Number MFCD00011401
    CAS 7778-77-0
    Health Hazard 1 H315-H319: Causes skin irritation. Causes serious eye irritation.
    Chemical Name or Material Potassium dihydrogen phosphate, For analysis ACS
    Grade ACS Grade

    H315-H319: Causes skin irritation. Causes serious eye irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit