Alfa Aesar™ Potassium ethoxide, 95%

Manufacturer: Alfa Aesar™ 1426318

Quantity 50g

 View more versions of this product

Catalog No. AA1426318

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 917-58-8
    Linear Formula KOCH2CH3
    Formula Weight 84.16
    MDL Number MFCD00054277
    UN Number UN3206
    Chemical Name or Material Potassium ethoxide

    Specifications

    Formula Weight 84.16
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H251-H314
    Self-heating: may catch fire.
    Causes severe skin burns and eye damage.
    Melting Point 250°C (dec.)
    Quantity 50g
    Sensitivity Air & Moisture Sensitive
    DOT Information Transport Hazard Class: 4.2; Packing Group: II; Proper Shipping Name: ALKALI METAL ALCOHOLATES, SELF-HEATING, CORROSIVE, N.O.S.
    TSCA TSCA
    Physical Form Powder
    UN Number UN3206
    Solubility Information Soluble in ethanol, ether
    Health Hazard 1 Danger
    Color Pale yellow to pale brown
    Linear Formula KOCH2CH3
    MDL Number MFCD00054277
    CAS 917-58-8
    Odor Alcohol-like
    Chemical Name or Material Potassium ethoxide
    Assay 95%

    GHS H Statement
    H251-H314
    Self-heating: may catch fire.
    Causes severe skin burns and eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit