Alfa Aesar™ Potassium ethoxide, 99+% (metals basis), 15% w/v in ethanol

Manufacturer: Alfa Aesar™ 39161AE

Quantity 100mL

 View more versions of this product

Catalog No. AA39161AE

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 917-58-8
    Linear Formula KOCH2CH3
    Formula Weight 84.16
    MDL Number MFCD00054277
    UN Number UN3274
    Chemical Name or Material Potassium ethoxide

    Specifications

    Formula Weight 84.16
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00054277
    UN Number UN3274
    Odor Alcohol-like
    TSCA TSCA
    CAS 917-58-8
    Health Hazard 1 Danger
    Quantity 100mL
    Sensitivity Air & Moisture Sensitive
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALCOHOLATES SOLUTION, N.O.S.
    Linear Formula KOCH2CH3
    Health Hazard 2 GHS H Statement
    H225-H314
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.
    Physical Form Liquid
    Color Colorless to pale yellow
    Concentration 15% w/v in ethanol
    Chemical Name or Material Potassium ethoxide
    Assay ≥99%, (metals basis)

    GHS H Statement
    H225-H314
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit