Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Potassium ethoxide, 99+% (metals basis), 15% w/v in ethanol

Click to view available options
Quantity:
25mL
917-58-8
C2H5KO
84.159
MFCD00054277
RPDAUEIUDPHABB-UHFFFAOYSA-N
potassium ethoxide,potassium ethanolate,potassium ethylate,ethanol, potassium salt 1:1,potassium ethoxide solution,ethanol, potassium salt,potassiumethoxide,potassium alcohol,etok,koet
23670592
potassium;ethanolate
CC[O-].[K+]
917-58-8
C2H5KO
84.159
MFCD00054277
RPDAUEIUDPHABB-UHFFFAOYSA-N
potassium ethoxide,potassium ethanolate,potassium ethylate,ethanol, potassium salt 1:1,potassium ethoxide solution,ethanol, potassium salt,potassiumethoxide,potassium alcohol,etok,koet
23670592
potassium;ethanolate
CC[O-].[K+]
Concentration or Composition (by Analyte or Components) 15% w/v in ethanol
Odor Alcohol-like
Sensitivity Air and moisture sensitive
Color Colorless to Yellow
Quantity 25mL
Linear Formula KOCH2CH3
UN Number UN3274
Solubility Information It reacts vigorously with water.
Formula Weight 84.16
Physical Form Liquid
Percent Purity ≥99%
Chemical Name or Material Potassium ethoxide
Assay (metals basis)
Show More Show Less

GHS H Statement
H225-H314
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.

P210-P233-P235-P235+P410-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H225-H314-H335-X

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALCOHOLATES SOLUTION, N.O.S.

EINECSNumber : 213-029-0

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon