Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium ferricyanide, 98%, pure, ACROS Organics™

Click to view available options
Quantity:
100g
500g
1kg
5kg
Packaging:
Glass bottle
Plastic Bottle
13746-66-2
C6FeK3N6
329.26
MFCD00011392
BYGOPQKDHGXNCD-UHFFFAOYSA-N
potassium ferricyanide, red prussiate, potassium ferricyanate, tripotassium hexacyanoferrate, potassium cyanoferrate, iron potassium cyanide, tripotassium ferriccyanide, tripotassium iron hexacyanide, tripotassium ferric hexacyanide, ccris 5559
26250
tripotassium;iron(3+);hexacyanide
[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[K+].[K+].[K+].[Fe+3]
13746-66-2
C6FeK3N6
329.26
MFCD00011392
BYGOPQKDHGXNCD-UHFFFAOYSA-N
potassium ferricyanide, red prussiate, potassium ferricyanate, tripotassium hexacyanoferrate, potassium cyanoferrate, iron potassium cyanide, tripotassium ferriccyanide, tripotassium iron hexacyanide, tripotassium ferric hexacyanide, ccris 5559
26250
tripotassium;iron(3+);hexacyanide
[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[K+].[K+].[K+].[Fe+3]
Indicator
Assay Percent Range 97.5% min. (Iodometry)
Linear Formula K3Fe(CN)6
Fieser 01,929; 02,345; 04,406; 05,554; 06,480; 07,300; 08,410; 09,385; 13,255
Merck Index 15, 7751
Solubility Information Solubility in water: 464g/L (20°C). Other solubilities: slightly soluble in alcohol
Formula Weight 329.26
Physical Form Crystalline Powder or Crystals
Percent Purity 98%
Grade Pure
Density 1.85
Packaging Glass bottle
Solubility Solubility in water: 464g/L (20°C). Other solubilities: slightly soluble in alcohol
Color Orange to Red
Quantity 100g
Chemical Name or Material Potassium ferricyanide, 98%
Show More Show Less

GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Contact with acids liberates very toxic gas.

GHS P Statement
Keep container tightly closed.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 237-323-3

RTECSNumber : LJ8225000

TSCA : TSCA