Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Ferrocyanide, Trihydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle

Chemical Identifiers

14459-95-1
K4Fe(CN)6·3H2O

High quality chemicals for laboratory and specialized industrial use

Formula Weight 422.41
Physical Form Crystals
Packaging Poly Bottle
Specific Gravity 1L = 1.85kg
Quantity 500g
Chemical Name or Material Potassium Ferrocyanide, Trihydrate
Grade Baker Analyzed™ ACS Reagent