Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Fluoride, 50% w/v, Neutralized, Reagents

Catalog No. 01334559
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Catalog No. Quantity
01-334-559 10 L
01-334-558 1 L
01-334-557 20 L
01-334-556 4 L
01-334-555 500 mL
5 options

Catalog No. 01-334-559

Supplier: Reagents Holdings Llc CP00230010F

Only null left
Add to Cart
Add to Cart

Potassium Fluoride, 50% w/v, Neutralized, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7789-23-3
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Boiling Point 102°C
Color Colorless
pH <8
Physical Form Liquid
Quantity 10 L
Percent Purity 50%
Grade Reagent
Packaging Cubitainer
Specific Gravity 1.354
Chemical Name or Material Potassium Fluoride, 50% w/v, Neutralized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.