Potassium fluoride, 99.9% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 4221706

Quantity 5g

 View more versions of this product

Catalog No. AA4221706

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 7789-23-3
    Physical Form Powder/Flake/Crystalline/Beads/Lump, Etc.
    Assay Percent Range 99.9% (metals basis)
    Molecular Formula FK
    Formula Weight 58.1
    MDL Number MFCD00011398
    Chemical Name or Material Potassium fluoride

    Specifications

    Physical Form Powder/Flake/Crystalline/Beads/Lump, Etc.
    Formula Weight 58.1
    Quantity 5g
    Color White
    DOT Information DOT Class: 6.1, Packaging Group: III
    Density 2.48g/cm3 at 20°C
    EINECS Number 232-151-5
    Assay Percent Range 99.9% (metals basis)
    Appearance White powder/flake/crystalline/beads
    MDL Number MFCD00011398
    Molecular Formula FK
    CAS 7789-23-3
    Vapor Pressure 0.13 hPa at 752°C
    Health Hazard 1 H301-H311-H331: Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled.
    Chemical Name or Material Potassium fluoride

    H301-H311-H331: Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit