Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium fluoride, 99+%, for analysis, ACROS Organics™

Click to view available options
Quantity:
250g
Packaging:
Plastic bottle
7789-23-3
FK
58.09
MFCD00011398
NROKBHXJSPEDAR-UHFFFAOYSA-M
potassium fluoride,potassiumfluoride,potassium fluoride kf,potassium fluorure,fluorure de potassium,potassium fluorure french,potassium fluoride on calcium fluoride,potassium fluoride, anhydrous,unii-9082wg1g3f,ccris 1568
522689
CHEBI:66872
potassium;fluoride
[F-].[K+]
7789-23-3
FK
58.09
MFCD00011398
NROKBHXJSPEDAR-UHFFFAOYSA-M
potassium fluoride,potassiumfluoride,potassium fluoride kf,potassium fluorure,fluorure de potassium,potassium fluorure french,potassium fluoride on calcium fluoride,potassium fluoride, anhydrous,unii-9082wg1g3f,ccris 1568
522689
CHEBI:66872
potassium;fluoride
[F-].[K+]
C-X bond formation reagent
Assay Percent Range 99+%
Linear Formula KF
Fieser 01,933; 02,346; 05,555; 06,481; 08,410; 10,325; 11,434; 12,406; 13,256; 15,167; 16,282
Merck Index 15, 7753
Solubility Information Solubility in water: 920g/L (18°C). Other solubilities: soluble in hydrofluoric acid,insoluble in alcohol unless water is present
Formula Weight 58.09
Physical Form Fine Crystalline Powder
Percent Purity 99+%
Grade Analysis
Cadmium (Cd) 5ppm max.
Chloride 50ppm max
Chlorine (Cl) 50ppm max.
Cobalt (Co) 5ppm max.
Copper (Cu) 5ppm max.
Density 2.48
Free Alkali 0.01% max. (K2CO3)
Free Acid 0.01% max.
Iron (Fe) 5ppm max.
Lead (Pb) 5ppm max.
Nickel (Ni) 5ppm max.
Packaging Plastic bottle
Potassium Fluosilicate 0.1% max.
Solubility Solubility in water: 920g/L (18°C). Other solubilities: soluble in hydrofluoric acid, insoluble in alcohol unless water is present
Sulfate 200ppm max.
Trace Analysis 5ppm max.
Boiling Point 1505.0°C
Color White
Melting Point 860.0°C
Quantity 250g
Chemical Name or Material Potassium fluoride, p.a.
Show More Show Less

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes serious eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air

GHS Signal Word: Danger

EINECSNumber : 232-151-5

RTECSNumber : TT0700000

TSCA : TSCA