Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium fluoride, ACS, 99% min

Manufacturer:  Alfa Aesar 4221636

Quantity 500g

 View more versions of this product

Catalog No. AA4221636


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7789-23-3
KF
MFCD00011398
3902818
Potassium fluoride
FK
58.1
UN1812
14,7632
ACS Grade

Specifications

7789-23-3
99% min
KF
MFCD00011398
3902818
Solubility in water at 21° (96g/100mL); Freely soluble in boiling water; Soluble in aqueous HF, liquid NH3; Insoluble in anhydrous alcohol
500g
≤99%
Powder
FK
58.1
UN1812
14,7632
2.48 g/mL
Potassium fluoride
ACS Grade

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H311-H331

GHS P Statement
P261-P280-P301+P310-P361-P304+P340-P311-P312-P363-P405-P501a

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: POTASSIUM FLUORIDE, SOLID

RTECSNumber : TT0700000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit