Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Fluoride, Anhydrous, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle

Chemical Identifiers

7789-23-3
FK
58.097
NROKBHXJSPEDAR-UHFFFAOYSA-M
522689
CHEBI:66872
potassium;fluoride
[F-].[K+]

High quality chemicals for laboratory and specialized industrial use

Formula Weight 58.1
Packaging Poly Bottle
Specific Gravity 1L = 2.48kg
Quantity 500g
Chemical Name or Material Potassium Fluoride, Anhydrous
Grade Baker Analyzed™ ACS Reagent