Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Fluoride, Anhydrous, Reagent, ACS, 99%, Spectrum™

Click to view available options
Quantity:
25 g
125 g
1 kg
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7789-23-3
FK
58.10
NROKBHXJSPEDAR-UHFFFAOYSA-M
potassium fluoride
[F-].[K+]
7789-23-3
FK
58.10
NROKBHXJSPEDAR-UHFFFAOYSA-M
potassium fluoride
[F-].[K+]

P1306, 7789-23-3, KF

Spectrum™ Potassium Fluoride, Anhydrous, Reagent, ACS is a source of the fluoride ion, and is the preferred reagent in the conversion process of chlorocarbons into fluorocarbons. As an ACS grade Reagent, Spectrum Chemical manufactured compounds are used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.

Grade ACS
CAS Max % 1
Percent Purity 99%
Heavy Metals (as Pb) 1e-005
Iron (Fe) 1e-005
Packaging Poly Pail
Sodium (Na) 0.002
Sulfate 5e-005
Quantity 12 kg