Alfa Aesar™ Potassium fluoride dihydrate, 98.5+%

Manufacturer: Alfa Aesar™ 1155514

Quantity 25g

 View more versions of this product

Catalog No. AA1155514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13455-21-5
    Formula Weight 94.13 (58.10 Anhydrous)
    MDL Number MFCD00149929
    UN Number UN1812
    Merck Index 14,7632
    Chemical Name or Material Potassium fluoride dihydrate

    Specifications

    Formula Weight 94.13 (58.10 Anhydrous)
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Odor Odorless
    Melting Point 41°C
    Health Hazard 2 GHS H Statement
    H301-H311-H331
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    Physical Form Crystalline
    MDL Number MFCD00149929
    CAS 13455-21-5
    UN Number UN1812
    Solubility Information Freely soluble in water
    TSCA TSCA
    Refractive Index 1.352
    Sensitivity Hygroscopic
    Merck Index 14,7632
    Quantity 25g
    Health Hazard 1 Danger
    Density 2.5g/mL
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: POTASSIUM FLUORIDE, SOLID
    Chemical Name or Material Potassium fluoride dihydrate
    Assay ≥98.5%

    GHS H Statement
    H301-H311-H331
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.

    GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit