Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium fluoride dihydrate, 98%, pure, ACROS Organics™

Click to view available options
Quantity:
5g
250g
Packaging:
Plastic bottle
13455-21-5
FH4KO2
94.127
MFCD00149929
MZFXHSQFQLKKFD-UHFFFAOYSA-M
potassium fluoride dihydrate,unii-95r1d12heh,potassium dihydrate fluoride,potassium fluoride dihydrate, 98.5+%,potassium fluoride dihydrate, puratronic,potassium fluoride, dihydrate,potassium fluoride kf , dihydrate,acmc-1byyf,ksc497c4l,potassiumfluoridedihydrate
23708904
potassium;fluoride;dihydrate
O.O.[F-].[K+]
13455-21-5
FH4KO2
94.127
MFCD00149929
MZFXHSQFQLKKFD-UHFFFAOYSA-M
potassium fluoride dihydrate,unii-95r1d12heh,potassium dihydrate fluoride,potassium fluoride dihydrate, 98.5+%,potassium fluoride dihydrate, puratronic,potassium fluoride, dihydrate,potassium fluoride kf , dihydrate,acmc-1byyf,ksc497c4l,potassiumfluoridedihydrate
23708904
potassium;fluoride;dihydrate
O.O.[F-].[K+]
C-X bond formation reagent
Assay Percent Range 98%
Linear Formula KF·2H2O
Fieser 01,933; 02,346; 05,555; 06,481; 08,410; 16,282
Formula Weight 94.13
Physical Form Crystals or Crystalline Powder
Percent Purity 19.65 to 20.70% (F)
Grade Pure
Density 2.45
Packaging Plastic bottle
Color Colorless to White
Quantity 250g
Chemical Name or Material Potassium fluoride dihydrate, 99%
Show More Show Less

GHS H Statement:
Toxic if swallowed.
Toxic if inhaled.
Toxic in contact with skin.

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

RTECSNumber : TT0705000