Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Fluoride, Dihydrate, Crystal, Reagent, 99-102%, Spectrum™

Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
6 x 500 g
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
13455-21-5
FH4KO2
94.13
MZFXHSQFQLKKFD-UHFFFAOYSA-M
potassium dihydrate fluoride
O.O.[F-].[K+]
13455-21-5
FH4KO2
94.13
MZFXHSQFQLKKFD-UHFFFAOYSA-M
potassium dihydrate fluoride
O.O.[F-].[K+]

P1310, 13455-21-5, KF•2H2O

Spectrum™ Potassium Fluoride, Dihydrate, Crystal, Reagent is a source of fluoride, it is the preferred reagent in the conversion process of chlorocarbons into fluorocarbons. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.

CAS Max % 1
Percent Purity 99 to 102%
Heavy Metals (as Pb) 2e-005
Insoluble Matter 0.0001
Packaging Poly Bottle
Sulfate 5e-005
Quantity 6 x 500 g