Alfa Aesar™ Potassium fluoride dihydrate, Puratronic™, 99.995% (metals basis)

Manufacturer: Alfa Aesar™ 1084109

Quantity 10g

 View more versions of this product

Catalog No. AA1084109

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13455-21-5
    Formula Weight 94.13 (58.10 Anhydrous)
    MDL Number MFCD00149929
    UN Number UN1812
    Merck Index 14,7632
    Chemical Name or Material Potassium fluoride dihydrate
    Grade Puratronic™

    Specifications

    Formula Weight 94.13 (58.10 Anhydrous)
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    TSCA TSCA
    MDL Number MFCD00149929
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H311-H331
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    Sensitivity Hygroscopic
    CAS 13455-21-5
    Merck Index 14,7632
    Density 2.5g/mL
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: POTASSIUM FLUORIDE, SOLID
    Odor Odorless
    Physical Form Powder
    Refractive Index 1.352
    UN Number UN1812
    Melting Point 41°C
    Quantity 10g
    Chemical Name or Material Potassium fluoride dihydrate
    Assay 99.995%, (metals basis)
    Grade Puratronic™

    GHS H Statement
    H301-H311-H331
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.

    GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit